Documenti fondamentali
Z176982
Sigma-Aldrich
Scienceware® serum-vial support
Holds 40 x 16-20 mm tubes
About This Item
Codice UNSPSC:
41122804
NACRES:
NB.35
Materiali
polypropylene
Caratteristiche
Holds 40 x 16-20 mm tubes
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F18513-2040
Lungh. × Largh. × Alt.
10 in. × 4 3/8 in. × 3 1/2 in.
Descrizione generale
Lightweight PP. Corrosion-proof. Fits most water baths for elevated temperature incubations. Top shelf height is 41mm (1⁵⁄₈").
Note legali
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)