Documenti fondamentali
Z170119
Sigma-Aldrich
Scienceware® glove box bare-hand port
iris port
Sinonimo/i:
humidity chambers, humidity control chambers, isolation chambers, nitrogen dry box
About This Item
Codice UNSPSC:
42132201
NACRES:
NB.35
Etichetta
E
Confezionamento
pack of 2 ea
Produttore/marchio commerciale
Bel-Art H50025-0020
Descrizione generale
The Scienceware glove box bare-hand port is for use when operations with gloves are difficult. Allows access to any glove box with a 73/8 in. I.D. port. Constructed of seven layers, 20 points of soft conforming silicone-latex rubber for an excellent seal. The bare-hand port is used with the Scienceware glove box, Z169943.
Note legali
Scienceware is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)