Z153931
Pouring rings for Duran® laboratory bottles
GL 45 neck thread, blue polypropylene
Sinonimo/i:
pouring ring
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Prodotti consigliati
Materiali
blue polypropylene
Produttore/marchio commerciale
DURAN®
Schott
Parametri
-40-140 °C temp. stability
Raccordi
GL 45 neck thread
Descrizione generale
The classic laboratory bottle GL threaded screw cap features the tried and tested cap plug sealing system ensuring a liquid tight seal. Available in four colours; blue, green, yellow and grey with matching pouring rings. Distinguishing media types is simplified and interchanging of screw caps and the risk of cross contamination is greatly reduced.
Caratteristiche e vantaggi
Optional pouring rings prevent drips when pouring from Duran bottles.
Note legali
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z153931-10EA | 4061832798110 |