Z107409
Aldrich® septum-inlet adapter with stopcock
Plain, joint: ST/NS 24/40, glass stopcock
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.35
Prodotti consigliati
Materiali
glass stopcock
Giunto
joint: ST/NS24/40
Categorie correlate
Descrizione generale
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Note legali
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z107409-1EA | 4061832109527 |