Documenti fondamentali
Z104647
Sigma-Aldrich
LDPE wash bottles
Narrow-mouth, capacity 500 mL
Sinonimo/i:
bottle lab, laboratory bottles, wash bottle
About This Item
Codice UNSPSC:
41103209
NACRES:
NB.17
Materiali
low-density polyethylene bottle
narrow-mouth bottle
natural bottle
polypropylene cap
Giunto
Joints threaded neck (38)
Caratteristiche
label no label
vented
with cap
Confezionamento
pack of 12 ea
Produttore/marchio commerciale
Bel-Art F11618-0016
Capacità
500 mL
Descrizione generale
Consist of one-piece cap and nozzle plus delivery tube. Deliver single drops or a needle-sharp stream. Tip may be cut to increase delivery amounts. Polyallomer cap.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)