Z100501
Kugelrohr glassware set
for standard capacity
About This Item
Codice UNSPSC:
41104812
NACRES:
NB.43
Descrizione generale
Set includes a straight tube with hose connection, 25 and 100 mL round-bottom oven flasks, and 25 and 100 mL single bulb receiver flasks with ST14/20 joints.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)