Documenti fondamentali
WHA77040001
Whatman® UNISEAL microplate seals
thin cold sealing film, adhesive backing, clear polypropylene
Sinonimo/i:
Z746827
About This Item
Codice UNSPSC:
41104900
NACRES:
NB.24
Materiali
clear polypropylene
Sterilità
non-sterile
Caratteristiche
thin cold sealing film, adhesive backing
Confezionamento
pkg of 100 ea
Produttore/marchio commerciale
Whatman 7704-0001
Whatman Article No. 28420886 (US reference)
Descrizione generale
Seals are used to control humidity and reduce evaporation of samples.
Caratteristiche e vantaggi
  • Prevent spills and contamination.
  • Cold seals are self-sticking with inert adhesive.
  • Heat seals are available in a clear polyester or aluminum foil.
  • Heat seals are for polypropylene microplates only and are applied with heat and pressure.
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)