Documenti fondamentali
WHA68962502
Filtri da siringa Whatman® GD/X
polyethersulfone membrane, pore size 0.2 μm, diam. 25 mm, sterile, pk of 50 ea
Sinonimo/i:
Whatman filter, Z286923, syringe filter, syringe filters
About This Item
Codice UNSPSC:
41104922
NACRES:
NB.24
Materiali
polyethersulfone membrane
polypropylene housing
Sterilità
sterile
Caratteristiche
glass fiber prefilter
Confezionamento
pk of 50 ea
Produttore/marchio commerciale
Whatman 6896-2502
Whatman Article No. 28420548 (US reference)
Diametro
25 mm
Volume
≤25 mL
Dimensione pori
0.2 μm
Raccordi
female Luer inlet (lock)
male Luer outlet
Descrizione generale
Sterile syringe filters with PES membrane and prefilter designed to support filtration of viscous, hard-to-filter samples with high solids content. The Whatman GD/X sterile, PES membrane syringe filters are designed to enable the filtration of viscous, hard-to-filter samples greater than 10 mL. Each unit contains a prefiltration stack of Whatman multigrade glass microfiber (GMF) and Glass fiber (GF/F) filters. The combination of this pre-filtration stack and filter housing design enables you to filter highly particular loaded samples. Cytiva′s GD/X sterile syringe filter is suitable for preparing viscous, dense, or otherwise hard-to-filter samples prior to a variety of analytical methods.
Caratteristiche e vantaggi
  • Reduces need for multiple syringe filters by processing three to seven times more sample volume than syringe filters without prefilters
  • Female luer lock for leak-free connections
  • High-efficiency Whatman GMF 150 and GF/F pre-filter media
  • Eliminates or greatly reduces the need to carry out separate prefiltration step for difficult-to-filter samples
Compatibilità
Suitable for filtering environmental and food analysis samples. Suitable for particulate laden sample preparation.
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)