Documenti fondamentali
WHA2200185
Whatman® 1PS phase separator filter papers
circles, diam. 185 mm, pack of 100
Sinonimo/i:
Z689785, phase separation filter papers, separation filter papers
About This Item
Codice UNSPSC:
41115727
NACRES:
NB.51
Materiali
circles
Confezionamento
pack of 100
Produttore/marchio commerciale
Whatman 2200-185
Whatman Article No. 28418827 (US reference)
Diametro
185 mm
Categorie correlate
Descrizione generale
Whatman 1PS Phase Separator from Cytiva′s business is a hydrophobic filter for routine solvent extraction. Within seconds it retains aqueous phases, while solvent phases flow through.
Hydrophobic solvent extraction paper
Solvent extraction can be time-consuming and messy when numerous separations are required. Cytiva′s Whatman 1PS Phase Separator is impregnated with stabilized silicone, creating a hydrophobic filter paper that can replace separatory funnel use in certain applications.
Rapid solvent extraction
Mixed phases are poured directly into the Whatman 1PS Phase Separator filter. The solvent phase flows through the paper quickly and cleanly, automatically stopping after the process is complete; all of the aqueous phase is retained. Droplets are automatically separated within seconds.
Caratteristiche e vantaggi
  • Impregnated with stabilized silicone to create a hydrophobic filter
  • Multiple separations can be processed in parallel
  • No special training required
  • Can replace a separatory funnel in certain applications
Altre note
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Note legali
Whatman is a registered trademark of Cytiva
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)