Documenti fondamentali
TEM-CFLF135CUUL
Sigma-Aldrich
Ultrathin Carbon Film Supported Copper London Finder LF135 Grids
box of 50
Sinonimo/i:
CARBON LONDON FINDER GRIDS - CU,LF135, ET 50/BX
About This Item
Codice UNSPSC:
41111761
NACRES:
MA.24
Confezionamento
box of 50
Descrizione generale
A 3-4 nm ultrathin film of pure carbon supported London finder (LF135) grids. Copper grid. Carbon film show better electrical and heat conduction in the electron beam. London finder grids are locator grids with reference/index on the grid.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)