Documenti fondamentali
SYNP150050C
Synthware synthesis vessel with lower joint and PTFE T-bore stopcock
50 mL, joint: ST/NS 24/40, porosity: coarse, with drip tip
Sinonimo/i:
Synthware P150050C, Solid phase peptide synthesis vessel
About This Item
Codice UNSPSC:
41100000
NACRES:
NB.51
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware P150050C
Capacità
50 mL
50 mL
Descrizione generale
Solid phase peptide synthesis vessels feature 2 mm PTFE stopcocks and fritted glass supports for resin beads during operation. The 24/40 inner drip joint can be assembled with other standard glassware. The vessel has a 8 mm hose connection for vacuum and the stopcock has an 8 mm O.D. sidearm for inert gas introduction. Vessels are supplied with PTFE-faced silicone lined Nylon cap.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)