SYNM211044
Synthware™ nitrogen/argon line manifold with PTFE stopcocks
port size 4, Hose Connections: left-right, stopcock size 4 mm
Sinonimo/i:
Synthware M211044, vacuum manifold, vacuum unit
About This Item
Codice UNSPSC:
40161501
NACRES:
NB.51
Prodotti consigliati
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware M211044
Dimensioni apertura
4
Misura rubinetto
4 mm
Descrizione generale
With 14/20 outer joint at the center to fit a rubber septum for keeping syringes under an inert environment cutting down on the number used. Main tube is 22 mm O.D. with both ends connections(10 mm). Delivery ports have 8 mm hose connections.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)