Key Documents
SYNF904200H
Synthware pear-shaped Schlenk flask with stopcock
capacity 200 mL, joint: ST/NS 24/40
Sinonimo/i:
Synthware F904200H, modified Schlenk reaction flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.51
Materiali
borosilicate glass flask
pear-shaped
single-neck
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F904200H
Capacità
200 mL
Descrizione generale
A suffix of G indicates a glass stopcock, H indicates a high-vacuum valve, no letter suffix indicates a PTFE Stopcock.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)