SYNF809600
Synthware™ Buchner funnel with perforated plate
600 mL, plate diam. 90 mm
Sinonimo/i:
Synthware F809600, filtering funnel
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.51
Prodotti consigliati
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F809600
Capacità
600 mL
600 mL
Diam. piatto
90 mm
Categorie correlate
Descrizione generale
With sealed-in perforated glass disc to support filter media. Pressure may be used but should not exceed 15 psi. This design solve the contamination problem with fritted disc and also the blindness problem with ceramic Buchner funnel.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)