SYNF601002F
Synthware™ Buchner funnel
2 mL, porosity: fine
Sinonimo/i:
Synthware F601002F, filtering funnel
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.51
Prodotti consigliati
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F601002F
Capacità
2 mL
2 mL
Categorie correlate
Descrizione generale
With sealed-in glass disc of the listed porosity. Not recommended for filtration where pressure would exceed 15psi. XC-extra coarse, C-Coarse, M-Medium, F-Fine porosity.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)