Documenti fondamentali
SYNF446500
Synthware three neck round bottom flask with vertical side necks
500 mL, center joint: ST/NS 29/42, side joint: ST/NS 24/40
Sinonimo/i:
Synthware F446500, 3-neck round flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.51
Materiali
borosilicate glass flask
round bottom
three-neck (vertical side necks)
Giunto
center joint: ST/NS29/42
side joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F446500
Capacità
500 mL
500 mL
Descrizione generale
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)