Key Documents
SYNF41241L
Synthware two neck round bottom flask with angled side neck
1000 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Sinonimo/i:
Synthware F41241L, 2-neck round flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.17
Materiali
borosilicate glass flask
round bottom
two-neck (angled side neck)
Giunto
center joint: ST/NS24/40
side joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F41241L
Capacità
1,000 mL
1000 mL
1000 mL
Descrizione generale
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)