Documenti fondamentali
SYNF301005
Synthware single neck round bottom flask
capacity 5 mL, joint: ST/NS 14/10
Sinonimo/i:
Synthware F301005, glass round flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.51
Materiali
borosilicate glass flask
round bottom
single-neck
Giunto
joint: ST/NS14/10
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F301005
Capacità
5 mL
Descrizione generale
Supplied with one holed compression caps, one Viton® O-rings and one PTFE - faced silicone rubber septum. For replacement caps, see RC112; for Viton® O-rings, see R014; for septa, see R1420.
Note legali
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)