SYNF283045C
Synthware™ fritted gas dispersion tube with 90 degree angle
disc diam. 30 mm, porosity: coarse, stem O.D. × L 8 mm × 150 mm
Sinonimo/i:
Synthware F282045M, glass gas dispersion tube
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.51
Prodotti consigliati
Materiali
colorless tube
glass tube
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F283045C
Diam. disco
30 mm
D.E. stelo × lungh.
8 mm × 150 mm
Categorie correlate
Descrizione generale
The 90° angle permits bubbles to be spread over a large area. Stem is 8 mm O.D. × 150 Length; XC stands for extra coarse porosity
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)