Documenti fondamentali
SYNF124125
Synthware single-neck flat-bottom flask
capacity 125 mL, joint: ST/NS 24/40
Sinonimo/i:
Synthware F124125, glass round flask
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.51
Materiali
borosilicate glass flask
flat bottom
single-neck
Giunto
joint: ST/NS24/40
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware F124125
Capacità
125 mL
Descrizione generale
Heavy wall single neck, flat bottom flask with an outer joint. Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)