SYNE114513X
Synthware™ extraction thimble
O.D. 45 mm, porosity: extra coarse, H 130 mm
Sinonimo/i:
Synthware E114513X, glass thimble
About This Item
Codice UNSPSC:
41104817
NACRES:
NB.51
Prodotti consigliati
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware E114513X
Altezza
130 mm
D.E.
45 mm
Categorie correlate
Descrizione generale
All glass extraction thimble permits viewing extraction process and subsequent weighing. C-Coarse, XC-Extra Coarse. Small thimble is used with C243445 and E103424; Medium thimble is used with C244500 and E104524; Large thimble is used with C245550 and E105550.
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
SYNE114513X-1EA | 4061832192239 |