Documenti fondamentali
SYNA201424
Synthware reducing adapter
top joint: ST/NS 14/20, bottom joint: ST/NS 24/40
Sinonimo/i:
Synthware A201424, glass reducing adapter, connecting adapter
About This Item
Codice UNSPSC:
41121815
NACRES:
NB.51
Giunto
bottom joint: ST/NS24/40
joint: ST/NS top14/20
Confezionamento
pkg of 1 ea
Produttore/marchio commerciale
Synthware A201424
Categorie correlate
Descrizione generale
Synthware reducing adapter is used for connecting dissimilar standard taper joint sizes. The bottom joint (24/40) is larger than the top joint (14/20).
Note legali
Synthware is a trademark of Kemtech America Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)