Documenti fondamentali
SLW1517/06
Pyrex® Media-Lab Bottles, with-out cap and pouring ring, with printed trace code
capacity 250 mL
Sinonimo/i:
Pyrex, SciLabware 1517/06
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Capacità:
250 mL
diam. × alt.:
70 mm × 138 mm
Materiali:
borosilicate glass bottle
round bottle
white graduation
Materiali
borosilicate glass bottle
round bottle
white graduation
Giunto
Joints threaded neck (GL45)
Caratteristiche
graduations
without cap
with printed trace code
without pouring ring
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
SciLabware 1517/06
Capacità
250 mL
diam. × alt.
70 mm × 138 mm
Descrizione generale
  • Bottles only. Supplied without cap or pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)