Documenti fondamentali
SLW1516/06D
Pyrex® Media-Lab Bottles, with cap and pouring ring, with printed trace code
capacity 250 mL
Sinonimo/i:
Pyrex, SciLabware 1516/06D
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
borosilicate glass bottle
polypropylene cap
round bottle
white graduation
Giunto
Joints threaded neck (GL45)
Sterilità
non-sterile
Caratteristiche
graduations
with cap
with pouring ring
with printed trace code
Confezionamento
pack of 10 ea
Produttore/marchio commerciale
SciLabware 1516/06D
Capacità
250 mL
diam. × alt.
70 mm × 138 mm
Descrizione generale
  • Bottles complete with polypropylene cap and drip-free pouring ring
  • Complies with ISO 4796
  • Printed with Trace Code for downloadable batch certificate
  • Batch certificate available to download from www.scilabware.com
  • Ideal for mixing and storage of culture medium
  • Sloping shoulders for smooth pouring action
  • White graduations and marking spot
  • Manufactured from temperature and chemically resistant borosilicate glass
  • For use up to 140 deg. C
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)