SLW1015/22D
Pyrex® Berzelius beakers, graduated, tall form with spout, with printed trace code
1000 mL
Sinonimo/i:
Pyrex, SciLabware 1015/22D
About This Item
Codice UNSPSC:
41121803
NACRES:
NB.43
Prodotti consigliati
Caratteristiche
graduated
Produttore/marchio commerciale
1015/22D
SciLabware
Altezza
180 mm
D.E.
95 mm
Capacità
1000 mL
Descrizione generale
- Complies with ISO 3819 and DIN 12331
- Manufactured from Pyrex® borosilicate glass
- Printed with Trace Code for downloadable batch certificate
- Batch certificate available to download from www.scilabware.com
- Tall form with spout
- White graduations and marking spot
Altre note
Browse all glass beakers
Note legali
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)