Documenti fondamentali
MT30046244
Mettler-Toledo SevenExcellence conductivity meter S700
S700 kit, meter only, AC/DC input 230 V AC, universal plug set
About This Item
Codice UNSPSC:
41115609
NACRES:
NB.61
Descrizione
Conductivity range 0.001 ?S/cm to 2000 ms/cm, various different units, defined by user (+/- 0.5%)
Resistivity range 0.01 ... 100.0 MOhm*cm
Produttore/marchio commerciale
Mettler-Toledo 30046244
Campo di misura della salinità
0-80 ppt
Campo di misura della temperatura
-30-130 °C, accuracy: ±0.1
Descrizione generale
Includes meter, a conductivity expansion unit and 2 blank expansion units
Mettler Toledo SevenExcellence conductivity S700 – intuitive touchscreen operation and high measurement performance combined with a complete security package for safe results and compliance. This single channel instrument is modularly expandable with additional measurement parameters at any time!
Professional, yet intuitive instrument with touchscreen operation, which can easily handle the most complex applications and meets stringent requirements in regulated markets.
  • Flexible due to various conductivity modes, units and temperature correction possibilities
  • Large 7 inch color display and intuitive menu guidance in 10 languages
  • Flexible method concept for high reproducibility and security
  • Extra security thanks to sophisticated user management and Intelligent Sensor Management (ISM®)
  • Integrated interfaces (USB, RS232 and Ethernet) for data exchange
  • Comprehensive service package including IQ/OQ/PQ
Included in every SevenExcellence:
uPlace electrode holder with a perfect vertical motion. It can be operated with one hand and used either freestanding or attached to the left or right side of the instrument.
LabX direct pH PC software for transferring real-time measurement data and archiving your results
Note legali
ISM is a registered trademark of Mettler-Toledo, Inc.
SevenExcellence is a trademark of Mettler-Toledo, Inc.
uPlace is a trademark of Mettler-Toledo, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)