DWKW242835-A
Wheaton® Dropping Bottle
white bottle, capacity 15 mL, case of 1000
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.17
Materiali
white bottle
Confezionamento
pkg of 1 packs
case of 1000
Produttore/marchio commerciale
WHEATON® W242835-A
Capacità
15 mL
Applicazioni
Opthalmic, IVD Kits, Other
Caratteristiche e vantaggi
- Bottle made of natural or white LDPE (Low Density Polyethylene
- White LDPE bottle helps protect UV light sensitive samples
- Dropper tip caps and tips sold separately
- Packaged in cases of 100 or 1000
Note legali
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
DWKW242835-A-1PK |