Documenti fondamentali
DWKW224682
WHEATON® amber sample vial with PTFE faced rubber lined cap packed in partitioned tray
glass, tube capacity (4 mL), screw cap
Sinonimo/i:
Sample Vial, W224682
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Materiali
amber borosilicate glass tube
flat bottom
glass
solid black phenolic cap
Sterilità
non-sterile
Caratteristiche
with cap
Confezionamento
case of 144 ea
Produttore/marchio commerciale
WHEATON® W224682
Capacità
(4 mL)
Diam. tubo
(15 mm)
Raccordi
neck (External Thread)
Descrizione generale
Amber Sample Vials in Lab File (With Caps Attached). Attached caps help maintain cleanliness. Supplied in partitioned trays to provide an easy way to store empty vials or to inventory samples.
  • Amber vials are manufactured from Wheaton 320 glass that conforms to USP Type I requirements to protect light-sensitive samples.
  • Screw caps are attached to vials.
  • It is suggested to use rubber lined caps for aqueous samples, and PTFE faced rubber lined caps for organic samples.
  • Vials and caps are autoclavable.
  • 2 mL vials fit the M-T Vial File, 228778.
  • 4 mL vials fit the M-T Vial File, 228780.
Note legali
Lab File is a registered trademark of Wheaton Industries, Inc.
Shorty Vial is a registered trademark of Wheaton Industries, Inc.
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)