DWK73802-15415
KIMBLE® solid cap with PTFE faced rubber liner
black phenolic cap, cap for 15-415
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.11
Prodotti consigliati
Materiali
PTFE/rubber liner
black phenolic cap
Sterilità
non-sterile
Caratteristiche
class A
Confezionamento
case of 500 ea
Produttore/marchio commerciale
Kimble® 73802-15415
Raccordi
cap for 15-415
Descrizione generale
Class A Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
- Specially formulated phenolic cap material
- Autoclavabl
- White rubber liners
- Excellent for general laboratory use
- Identified in ASTM Specification E982, Class A requirement
Caratteristiche e vantaggi
- Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
- PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
- Excellent for general laboratory use
- Identified in ASTM Specification E982, Class A requirements
Note legali
KIMBLE is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)