DWK11-5200-S
Wheaton® Microliter Vial
About This Item
Codice UNSPSC:
41122001
NACRES:
NB.11
Prodotti consigliati
Materiali
clear vial (Silanized, with Marking Patch)
Confezionamento
pkg of 100 ea
Produttore/marchio commerciale
WHEATON® 11-5200-S
D × alt.
12 mm × 32 mm
Dimens. tappo
11 mm
Volume
2.0 mL
Applicazioni
Chromatography
Caratteristiche e vantaggi
- Double ring finish for added closure performance.
- Approved for LC & GC.
- Where noted, Certified Class 10,000 clean room packed enhances data integrity.
Note legali
WHEATON is a registered trademark of Wheaton Industries, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
DWK11-5200-S-100EA |