Documenti fondamentali
CLS70980500
Pyrex® VISTA Erlenmeyer flask, narrow-mouth
capacity 500 mL
Sinonimo/i:
conical flask, conical flasks, erlenmeyer flasks, flask erlenmeyer, Corning® 70980
About This Item
Codice UNSPSC:
41121804
NACRES:
NB.43
Materiali
borosilicate glass
narrow-mouth
Sterilità
non-sterile
Caratteristiche
cap: no
Confezionamento
case of 12
pack of 6
Produttore/marchio commerciale
Corning 70980-500
Capacità
500 mL
Descrizione generale
These narrow mouth Erlenmeyer flasks have uniform wall thickness which provide the proper balance between mechanical strength and thermal shock resistance. Approximate graduations are in durable blue enamel. An extra large marking space is also provided.
Altre note
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
VISTA is a trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)