CLS44462L
Pyrex® shaker flask with extra-deep baffles
capacity 2,000 mL
Sinonimo/i:
Corning® 4446
About This Item
Codice UNSPSC:
41121804
Prodotti consigliati
Materiali
Delong
borosilicate glass
Sterilità
non-sterile
Caratteristiche
cap: no
closure type Closure cap
Confezionamento
case of 3
Produttore/marchio commerciale
Corning 4446-2L
Capacità
2,000 mL
Descrizione generale
Delong style neck, with three extra-deep baffles, to enhance gas transfer when used with rotary or reciprocating shakers. Long neck reduces splashing and is designed for two-position polypropylene closures. Closures are sold separately.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)