Documenti fondamentali
CLS2962500
Pyrex® hydrometer jar
capacity 500 mL
Sinonimo/i:
Corning® 2962
About This Item
Codice UNSPSC:
41113024
NACRES:
NB.43
Produttore/marchio commerciale
Corning 2962-500
D.E. × alt.
50 mm × 390 mm
Capacità
500 mL
Descrizione generale
Breakage is minimized by the heavy wall construction. The large hexagonal base, sealed to the cylinder body, increases stability. Meets ASTM specifications D-287 and E-100.
Note legali
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)