BR704010
BRAND® bottle for dispensers
capacity 2500 mL, with Ethyl-Acrylate coat
Sinonimo/i:
Accessories, Bottle-top Dispenser, Bottles, Dispensette®
About This Item
Prodotti consigliati
Materiali
amber bottle
narrow-mouth bottle
polypropylene cap
round bottle
soda-lime glass bottle (with Ethyl-Acrylate coat)
Giunto
Joints threaded neck (GL45)
Caratteristiche
with cap
with Ethyl-Acrylate coat
with PP screw cap GL45
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 704010
Capacità
2500 mL
diam. × alt.
140 mm × 300 mm
Note legali
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR704010-1EA | 4061831858334 |