Documenti fondamentali
BR6783
BRAND® replacement amber inspection windows for Titrette®
Sinonimo/i:
Accessories, Bottle-top Burette, Burettes, Inspection window, Titrette®
About This Item
Codice UNSPSC:
41121500
NACRES:
NB.43
Confezionamento
pack of 2 SETS
Produttore/marchio commerciale
BRAND 6783
Descrizione generale
For protection of light-sensitive media, you can replace the clear inspection windows with the amber colored windows (included).
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Titrette is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)