BR65975
BRAND® dessicator plate
for nominal size 200 mm, O.D. 190 mm, porcelain
Sinonimo/i:
Accessories, Desiccator plate, Desiccators
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Materiali
porcelain
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 65975
D.E.
190 mm
Compatibilità
suitable for (glass dessicators)
Categorie correlate
Descrizione generale
Our porcelain plates for glass desiccators and polypropylene (PP) plates for PC/PP desiccators are essential components designed to enhance the functionality and efficiency of your desiccation processes. Both porcelain and PP plates are designed to optimize the performance of your desiccators, ensuring that your samples remain protected from moisture and environmental contaminants.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR65975-1EA | 4061831857771 |