BR65731
BRAND® desiccator ′Novus′ lid with socket NS 24/29
nominal diameter × O.D. × H 150 mm × 215 mm × 78 mm, joint: 24/29
Sinonimo/i:
Accessories, Lids
About This Item
Codice UNSPSC:
41104510
NACRES:
NB.43
Prodotti consigliati
Giunto
joint: 24/29
Caratteristiche
lid with socket
Produttore/marchio commerciale
BRAND 65731
Diametro nominale × D.E. × alt.
150 mm × 215 mm × 78 mm
Categorie correlate
Descrizione generale
Crafted with high-quality materials, this lid features a robust knob for easy handling and secure sealing. Specifically designed for use with BRAND® glass desiccators, providing a perfect fit. Specifically designed for use with BRAND® glass desiccators, providing a perfect fit.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR65731-1EA | 4061831857702 |