BR472700
BRAND® glass staining trough, Hellendahl extended
soda-lime glass
Sinonimo/i:
Microscopy, Staining, Staining Accessories
About This Item
Codice UNSPSC:
41122800
NACRES:
MA.01
Prodotti consigliati
Materiali
soda-lime glass
Produttore/marchio commerciale
BRAND 472700
Lungh. × largh.
76 mm × 26 mm , holds 16 slides
Descrizione generale
The BRAND® Glass Staining Trough, Hellendahl Extended, is designed for the efficient staining of up to 16 slides, making it an essential tool for laboratory applications. Made from durable soda-lime glass, this trough features dimensions of 76 x 26 mm, providing ample space for multiple slides while ensuring optimal organization. The Hellendahl design allows for effective staining and easy access to each slide, enhancing workflow efficiency. This reliable staining trough is ideal for histology and microscopy procedures, making it a valuable addition to any laboratory setup.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)