Documenti fondamentali
BR43205
BRAND® density bottle, Gay-Lussac pattern, uncalibrated
capacity 5 mL, borosilicate glass
Sinonimo/i:
Gay-Lussac
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Materiali
borosilicate glass
glass stopper
narrow-mouth bottle
round bottle (Gay-Lussac)
white bottle
Giunto
joint ST/NS neck10/19
Caratteristiche
with cap
Confezionamento
pack of 2 ea
Produttore/marchio commerciale
BRAND 43205
Capacità
5 mL
Descrizione generale
DIN ISO 3507, type Gay-Lussac. Stopper NS 10/19 with capillary. Top of stopper ground and polished. Nominal capacity printed on the bottom.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Visita l’Archivio dei documenti
Protocolli
Density bottles are mainly used to determine the density of liquids of moderate viscosity. They are not volumetric instruments, however, they are calibrated 'to contain' as in the case of volumetric flasks.