Documenti fondamentali
BR16515
BRAND® burette clamp
Zinc alloy, nickel-plated for 1 burette, clamp PVC-coated
Sinonimo/i:
Accessories, Burette clamp, Burettes
About This Item
Codice UNSPSC:
41122413
NACRES:
NB.01
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 16515
Descrizione generale
Burettes up to 50 ml can be mounted in seconds. PVC-coated clamps provide a secure grip. Comfortable height adjustment. Graduations remain visible. Very sturdy design, easy to operate. Integral support connector.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)