Key Documents
BR156640
BRAND® tray stackable
size 225 x 180 x 45 mm
Sinonimo/i:
Photographoc trays, Trays
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.51
Materiali
polypropylene
white
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 156640
Misura
225 x 180 x 45 mm
Descrizione generale
The BRAND® stackable tray is made from durable white polypropylene (PP), designed for versatile use in laboratory and industrial settings. Its ribbed bottom enhances stability, preventing contents from shifting during transport or storage. The reinforced rim adds strength, while the slightly sloping walls facilitate easy access and pouring. This stackable design allows for efficient storage, maximizing space when not in use. Ideal for organizing and handling various materials, this tray combines practicality with durability, making it an essential tool for any workspace.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)
Lot/Batch Number
Ci dispiace, ma al momento non ci sono COA disponibili online per questo prodotto.
Visita l’Archivio dei documenti
I clienti hanno visto anche
Slide 1 of 6
1 of 6
BRAND® tray stackable size 840 x 645 x 160 mm
BR156650
BRAND® tray stackable
Autoclave pans capacity 1.8 L
Z360376
Autoclave pans
Autoclave pans capacity 5 L
Z360406
Autoclave pans