Documenti fondamentali
BR148502
BRAND® funnel holder
Holds 2 funnels
Sinonimo/i:
Accessories, Funnels
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.35
Materiali
polypropylene
Caratteristiche
Holds 2 funnels
Confezionamento
pack of 5 ea
Produttore/marchio commerciale
BRAND 148502
Descrizione generale
PP. To support 1 or 2 funnels with top outer-Ø of 50 to 120 mm. Easily mounted on supports rods with an outer-Ø 8-14 mm with a locking nut
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)