BR144610
BRAND® BISTABIL® ground glass stopper, conical joint
borosilicate glass 3.3, joint: ST/NS 10/19, (hollow, hexagonal grip)
About This Item
Codice UNSPSC:
41121800
NACRES:
NB.43
Prodotti consigliati
Materiali
borosilicate glass 3.3
Giunto
joint: ST/NS10/19
Produttore/marchio commerciale
BRAND 144610
Categorie correlate
Descrizione generale
Conical ground joint stopper made from Duran®. Hollow with drip tip and hexagonal grip. Meets DIN 12 252.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Bistabil is a registered trademark of Lenz GmbH
DURAN is a registered trademark of DWK Life Sciences
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)