BR140004
BRAND® separating funnel with screw cap
size 125 mL, polypropylene
Sinonimo/i:
Funnels, Separating funnels
About This Item
Codice UNSPSC:
41121809
NACRES:
NB.43
Prodotti consigliati
Materiali
conical PTFE stopcock
polypropylene
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
BRAND 140004
Misura
125 mL
Categorie correlate
Descrizione generale
Polypropylene, high clarity for observation of contents. With polypropylene screw cap and special conical PTFE stopcock with retention device. Autoclavable (121 °C).
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)