Documenti fondamentali
BR135825
BRAND® stirring rod
AR-Glass, L × D 300 mm × 8 mm
Sinonimo/i:
Stirring rods
About This Item
Codice UNSPSC:
41121802
NACRES:
NB.72
Materiali
AR-Glass
Descrizione
fused closely at both ends
Confezionamento
pack of 50 ea
Produttore/marchio commerciale
BRAND 135825
L × D
300 mm × 8 mm
Descrizione generale
The BRAND® stirring rod is a glass tube that is closed on both ends, providing a reliable and durable tool for mixing and stirring various solutions in laboratory settings. Its smooth surface ensures easy cleaning and maintenance, while the closed design prevents contamination.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)