Documenti fondamentali
BR114940
BRAND® mini cooler
PC, for 12 tubes
Sinonimo/i:
Accessories, Microcentrifuge Tubes, Mini cooler, Sample Storage, Tubes
About This Item
Codice UNSPSC:
41103025
NACRES:
NB.22
Materiali
PC
Descrizione
bench temp.maintained -70°C time 45 min.
Caratteristiche
for 12 tubes
Produttore/marchio commerciale
BRAND 114940
Categorie correlate
Descrizione generale
PC. Mini coolers are designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench. Durable polycarbonate filled with non-toxic gel. Mini coolers hold twelve 0.5 ml to 2.0 ml tubes.
Note legali
BRAND is a registered trademark of BRAND GMBH + CO KG
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)