Documenti fondamentali
BAH131760612
SP Bel-Art® utility bags
width × H × thickness 6 in. (152 mm) × 12 in. (305 mm) × 1.5 mil (0.038 mm), clear, pack of 100 ea
Sinonimo/i:
clear bag, reusable bag, sample bag
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.35
Materiali
clear
Confezionamento
pack of 100 ea
Produttore/marchio commerciale
Bel-Art H13176-0612
width × H × thickness
6 in. (152 mm) × 12 in. (305 mm) × 1.5 mil (0.038 mm)
Descrizione generale
Reusable, transparent bags are ideal for shipping or storage and are especially useful as sample bags.
Caratteristiche e vantaggi
  • FDA grade polyethylene, safe for food, drug and cosmetic use
  • Latex free
  • Not Autoclavable
  • H13176 lightweight bags are 0.038 mm (1.5 mil) thick, H13178 heavyweight bags are 0.1016 mm (4 mil) thick
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)