Merck
Tutte le immagini(7)
Documenti fondamentali
BAF371220060
SP Bel-Art® Spinbar® magnetic stir bar
polygon white PTFE-coated, L × diam. 2.375 in. (60 mm) × 0.375 in. (10 mm)
Sinonimo/i:
Spinbar® magnetic stir bar
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.51
Materiali
polygon white PTFE-coated
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F37122-0060
L × diam.
2.375 in. (60 mm) × 0.375 in. (10 mm)
Descrizione generale
  • Spinbar® magnetic stir bar is used in conjunction with a magnetic stirring plate for uniform, consistent, and thorough mixing of chemical solutions.
  • Alnico magnet encapsulated in durable and inert FDA grade polytetrafluoroethylene (PTFE) low friction coating.
  • Multi-faceted-shaped bars provide a greater surface area and added turbulence for more vigorous, thorough mixing.
  • Autoclave safe.
These stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
Caratteristiche e vantaggi
  • Center molded pivot ring minimizes the contact area between the bar and the vessel; reduces friction and chattering
  • Marring of plastic containers is diminished
  • Alnico magnet encapsulated in FDA Grade, USP Class VI PTFE® brand PTFE Coating – Superior Chemical Compatibility
  • Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
  • Autoclave safe
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Spinbar is a registered trademark of SP Scienceware, a division of SP
Teflon is a registered trademark of E. I. du Pont de Nemours and Company
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)