Documenti fondamentali
BAF196140000
SP Bel-Art® Wye (Y) Tubing Connectors
tubing I.D. 0.25 in. (6 mm)
Sinonimo/i:
connectors, tubing, tubing coupler
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.43
Confezionamento
pack of 12 ea
Produttore/marchio commerciale
Bel-Art F19614-0000
Lungh. × largh.
1.3125 in. (33.3 mm) × 2.0625 in. (52.8 mm)
D.I. tubo
0.25 in. (6 mm)
Descrizione generale
Use these fittings to ensure a tight, secure connection between lengths of tubing having the same inner diameter (I.D.).
Caratteristiche e vantaggi
  • Multiple barbs per arm provide additional contact between the fitting and the tubing for extra gripping power
  • Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases
  • FDA-grade for direct contact with food, drugs, and cosmetics
  • Autoclavable
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)