Key Documents
BAF171300024
SP Bel-Art® Pipette Jar for Cleanware Pipette Rinsing System
diam. × H 15.2 cm (6 in.) × 61 cm (24 in.)
About This Item
Codice UNSPSC:
41122800
NACRES:
NB.01
Confezionamento
pack of 1 ea
Produttore/marchio commerciale
Bel-Art F17130-0024
diam. × alt.
15.2 cm (6 in.) × 61 cm (24 in.)
Descrizione generale
Store and soak pipettes in these leakproof, molded polyethylene Pipette Jars.
Caratteristiche e vantaggi
  • Withstands all common cleaning solutions at room temperature, including sulfuric acid/dichromate
Note legali
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Scegli una delle versioni più recenti:
Certificati d'analisi (COA)